Please login to your online account to display your discounted pricing

anti-LAMP-1/CD107a, Clone: 6E2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225155

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225155

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Isotype IgG1
Quantity 0.1mL
Antigen LAMP-1/CD107a
Immunogen Full length human LAMP1 [UniProt No. P11279]
Format Affinity Purified
Clone 6.00E+02
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Gene Accession No. P11279
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Human, Bovine, Porcine, Mouse and Rat. Other species have not been tested.
Conjugate Unlabeled
Formulation PBS
Gene ID (Entrez) 3916
Concentration 1.6mg/mL
Host Species Mouse
Regulatory Status RUO