Please login to your online account to display your discounted pricing

anti-LAMP-1/CD107a, Clone: 6E2, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone 6.00E+02
Host Species Mouse
Isotype IgG1
Immunogen Full length human LAMP1 [UniProt No. P11279]
Antigen LAMP-1/CD107a
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP225155SS

 
novus biologicals canada
NBP225155SS
Unlabeled 0.025mL Each for $109.00

NBP225155

 
novus biologicals canada
NBP225155
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Clone 6.00E+02
Host Species Mouse
Isotype IgG1
Immunogen Full length human LAMP1 [UniProt No. P11279]
Antigen LAMP-1/CD107a
Monoclonal or Polyclonal Monoclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity Human, Bovine, Porcine, Mouse and Rat. Other species have not been tested.
Gene Accession No. P11279
Gene ID (Entrez) 3916
Formulation PBS
Concentration 1.6mg/mL