Please login to your online account to display your discounted pricing

anti-Lambda light chain, Clone: LcN-2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP222386

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP222386

  • / Each
 

Description & Specifications

Specifications

Antigen Lambda light chain
Regulatory Status RUO
Immunogen Human IgG was used as immunogen to generate the Lambda LcN-2 antibody.
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Format Purified
Isotype κ
Formulation PBS with 0.05% BSA.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3536
Concentration 0.2mg/mL
Conjugate Unlabeled
Host Species Mouse
Quantity 0.1mg
Clone LcN-2
Cross Reactivity Human