Please login to your online account to display your discounted pricing

anti-Lambda light chain, Clone: LcN-2, Novus Biologicals™

$362.00 - $362.00

Specifications

Clone LcN-2
Host Species Mouse
Isotype κ
Immunogen Human IgG was used as immunogen to generate the Lambda LcN-2 antibody.
Antigen Lambda light chain
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP222386

 
novus biologicals canada
NBP222386
Unlabeled 0.1mg Each for $362.00
Description & Specifications

Specifications

Clone LcN-2
Host Species Mouse
Isotype κ
Immunogen Human IgG was used as immunogen to generate the Lambda LcN-2 antibody.
Antigen Lambda light chain
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene ID (Entrez) 3536
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL