Please login to your online account to display your discounted pricing

anti-Lambda light chain, Clone: LcN-2, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP2223861MG

 View all options for this product


For Research Use Only

Catalog No. NBP2223861


Description & Specifications


Antigen Lambda light chain
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Clone LcN-2
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen Human IgG was used as immunogen to generate the Lambda LcN-2 antibody.
Isotype κ
Quantity 1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3536