Please login to your online account to display your discounted pricing

anti-Lactoferrin, Clone: 2B8, Novus Biologicals™


Manufacturer: novus biologicals canada  NB6001529

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB6001529

  • / Each
 

Description & Specifications

Specifications

Quantity 0.1mg
Format Affinity Purified
Formulation Constituents: PBS. pH 7.4
Isotype IgG1, κ
Clone 2B8
Antigen Lactoferrin
Applications Sandwich ELISA
Applications Western Blotting
Conjugate Unlabeled
Primary or Secondary Primary
Cross Reactivity Human
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4057
Test Specificity Does not cross-react with human transferrin or other serum proteins. Cross-reacts with Human. Not yet tested in other species.
Host Species Mouse
Regulatory Status RUO
Immunogen Full length native protein (purified) (Human).