Please login to your online account to display your discounted pricing

anti-Kallikrein 3/PSA, Clone: A67-B/E3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232992

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232992

  • / Each
 

Description & Specifications

Specifications

Antigen Kallikrein 3/PSA
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone A67-B/E3
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P07288
Host Species Mouse
Immunogen PSA from human sperm plasma
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 354