Please login to your online account to display your discounted pricing

anti-Kallikrein 3/PSA, Clone: A67-B/E3, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone A67-B/E3
Host Species Mouse
Isotype κ
Immunogen PSA from human sperm plasma
Antigen Kallikrein 3/PSA
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232992

 
novus biologicals canada
NBP232992
Unlabeled 0.5mL Each for $321.00

NBP2329921M

 
novus biologicals canada
NBP2329921ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone A67-B/E3
Host Species Mouse
Isotype κ
Immunogen PSA from human sperm plasma
Antigen Kallikrein 3/PSA
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P07288
Gene ID (Entrez) 354
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL