Please login to your online account to display your discounted pricing

anti-Kallikrein 3/PSA, Clone: 5A11E9, Novus Biologicals™

$139.00 - $309.05

Specifications

Clone 5A11E9
Host Species Murine
Isotype IgG1
Immunogen Purified recombinant fragment of Prostate Specific Antigen (amino acids 26-251) expressed in &italic_begin;E. coli&italic_end;. [UniProt No. P07288]
Antigen Kallikrein 3/PSA
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP151541

 
novus biologicals canada
NBP151541
Unlabeled 0.1mL Each for $309.05

NBP151541SS

 
novus biologicals canada
NBP151541SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Clone 5A11E9
Host Species Murine
Isotype IgG1
Immunogen Purified recombinant fragment of Prostate Specific Antigen (amino acids 26-251) expressed in &italic_begin;E. coli&italic_end;. [UniProt No. P07288]
Antigen Kallikrein 3/PSA
Monoclonal or Polyclonal Monoclonal
Format Ascites
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene Accession No. P07288
Gene ID (Entrez) 354
Concentration 1.0mg/mL