Please login to your online account to display your discounted pricing

anti-Integrin beta 4/CD104, Clone: UM-A9, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP2328131ML

 View all options for this product


For Research Use Only

Catalog No. NBP2328131M


Description & Specifications


Antigen Integrin beta 4/CD104
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Clone UM-A9
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P16144
Host Species Mouse
Immunogen Human squamous cell carcinoma (UM-SCC1)
Isotype κ
Quantity 1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3691