Please login to your online account to display your discounted pricing

anti-Integrin beta 4/CD104, Clone: UM-A9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232813

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232813

  • / Each
 

Description & Specifications

Specifications

Format Purified
Antigen Integrin beta 4/CD104
Cross Reactivity Human
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Human squamous cell carcinoma (UM-SCC1)
Isotype κ
Clone UM-A9
Concentration 0.2mg/mL
Formulation PBS with 0.05% BSA.
Primary or Secondary Primary
Gene ID (Entrez) 3691
Conjugate Unlabeled
Gene Accession No. P16144
Monoclonal or Polyclonal Monoclonal
Quantity 0.5mL
Regulatory Status RUO