Please login to your online account to display your discounted pricing

anti-Integrin beta 4/CD104, Clone: UM-A9, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone UM-A9
Host Species Mouse
Isotype κ
Immunogen Human squamous cell carcinoma (UM-SCC1)
Antigen Integrin beta 4/CD104
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232813

 
novus biologicals canada
NBP232813
Unlabeled 0.5mL Each for $321.00

NBP2328131M

 
novus biologicals canada
NBP2328131ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone UM-A9
Host Species Mouse
Isotype κ
Immunogen Human squamous cell carcinoma (UM-SCC1)
Antigen Integrin beta 4/CD104
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P16144
Gene ID (Entrez) 3691
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL