Please login to your online account to display your discounted pricing

anti-Integrin alpha L/CD11a, Clone: CRIS-3, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP232923

 View all options for this product


For Research Use Only

Catalog No. NBP232923


Description & Specifications


Antigen Integrin alpha L/CD11a
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Clone CRIS-3
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P20701
Host Species Mouse
Immunogen Stimulated human leukocytes
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3683