Please login to your online account to display your discounted pricing

anti-Integrin alpha L/CD11a, Clone: CRIS-3, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone CRIS-3
Host Species Mouse
Isotype κ
Immunogen Stimulated human leukocytes
Antigen Integrin alpha L/CD11a
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232923

 
novus biologicals canada
NBP232923
Unlabeled 0.5mL Each for $321.00

NBP2329231M

 
novus biologicals canada
NBP2329231ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone CRIS-3
Host Species Mouse
Isotype κ
Immunogen Stimulated human leukocytes
Antigen Integrin alpha L/CD11a
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Gene Accession No. P20701
Gene ID (Entrez) 3683
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL