Please login to your online account to display your discounted pricing

anti-Integrin alpha 2b/CD41, Clone: MEM-06, Novus Biologicals™


Manufacturer: novus biologicals canada  NB500459

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB500459

  • / Each
 

Description & Specifications

Specifications

Antigen Integrin alpha 2b/CD41
Format Purified
Host Species Mouse
Immunogen Tissue/cell preparation (Human). (Leucocytes of patient suffering from LGL-type leukaemia).
Isotype IgG1
Quantity 0.1mg
Regulatory Status RUO
Clone MEM-06
Cross Reactivity Human
Formulation PBS, pH 7.4
Applications Flow Cytometry
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3674
Concentration 1mg/mL
Conjugate Unlabeled