Please login to your online account to display your discounted pricing

anti-Integrin alpha 2b/CD41, Clone: MEM-06, Novus Biologicals™

$110.17 - $307.00

Specifications

Clone MEM-06
Host Species Murine
Isotype IgG1
Immunogen Tissue/cell preparation (Human). (Leucocytes of patient suffering from LGL-type leukaemia).
Antigen Integrin alpha 2b/CD41
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB500459

 
novus biologicals canada
NB500459
Unlabeled 0.1mg Each for $307.00

NB500459SS

 
novus biologicals canada
NB500459SS
Unlabeled 0.02mg Each for $110.17
Description & Specifications

Specifications

Clone MEM-06
Host Species Murine
Isotype IgG1
Immunogen Tissue/cell preparation (Human). (Leucocytes of patient suffering from LGL-type leukaemia).
Antigen Integrin alpha 2b/CD41
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Gene ID (Entrez) 3674
Formulation PBS, pH 7.4
Concentration 1mg/mL