Please login to your online account to display your discounted pricing

anti-Integrin alpha 2b/CD41, Clone: MEM-06, Novus Biologicals™


Manufacturer: novus biologicals canada  NB500459SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB500459SS

  • / Each
 

Description & Specifications

Specifications

Format Purified
Regulatory Status RUO
Clone MEM-06
Isotype IgG1
Antigen Integrin alpha 2b/CD41
Cross Reactivity Human
Immunogen Tissue/cell preparation (Human). (Leucocytes of patient suffering from LGL-type leukaemia).
Quantity 0.02mg
Formulation PBS, pH 7.4
Applications Flow Cytometry
Concentration 1mg/mL
Conjugate Unlabeled
Primary or Secondary Primary
Gene ID (Entrez) 3674
Monoclonal or Polyclonal Monoclonal
Host Species Mouse