Please login to your online account to display your discounted pricing

anti-Insulin/Proinsulin, Clone: D3E7, Novus Biologicals™

$110.17 - $219.00

Specifications

Clone D3E7
Host Species Murine
Isotype IgG1, κ
Immunogen Kd for this is 6.3 x 108M.This is specific for both insulin and proinsulin
Antigen Insulin/Proinsulin
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB200435

 
novus biologicals canada
NB200435
Unlabeled 0.2mg Each for $219.00

NB200435SS

 
novus biologicals canada
NB200435SS
Unlabeled 0.04mg Each for $110.17
Description & Specifications

Specifications

Clone D3E7
Host Species Murine
Isotype IgG1, κ
Immunogen Kd for this is 6.3 x 108M.This is specific for both insulin and proinsulin
Antigen Insulin/Proinsulin
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Enzyme Immunoassay
Test Specificity This is specific for both insulin and proinsulin. Not yet tested in other species.
Gene ID (Entrez) 3630
Formulation PBS, pH 7.4