Please login to your online account to display your discounted pricing

anti-IL1RL2, Clone: 18D6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP233300

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP233300

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q9HB29
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Gene ID (Entrez) 8808
Formulation PBS
Concentration 1.0mg/mL
Cross Reactivity Human
Host Species Mouse
Isotype κ
Format Purified
Regulatory Status RUO
Quantity 0.1mg
Clone 18D6
Immunogen Partial recombinant human IL1RL2 (between amino acids 1-250) [UniProt Q9HB29]
Antigen IL1RL2
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting