Please login to your online account to display your discounted pricing

anti-IL1RL2, Clone: 18D6, Novus Biologicals™

$109.00 - $307.00

Specifications

Clone 18D6
Host Species Mouse
Isotype κ
Immunogen Partial recombinant human IL1RL2 (between amino acids 1-250) [UniProt Q9HB29]
Antigen IL1RL2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP233300

 
novus biologicals canada
NBP233300
Unlabeled 0.1mg Each for $307.00

NBP233300SS

 
novus biologicals canada
NBP233300SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Clone 18D6
Host Species Mouse
Isotype κ
Immunogen Partial recombinant human IL1RL2 (between amino acids 1-250) [UniProt Q9HB29]
Antigen IL1RL2
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene Accession No. Q9HB29
Gene ID (Entrez) 8808
Formulation PBS
Concentration 1.0mg/mL