Please login to your online account to display your discounted pricing

anti-IL1RL2, Clone: 15G9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP233299SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP233299SS

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Gene ID (Entrez) 8808
Gene Accession No. Q9HB29
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Concentration 1.0mg/mL
Antigen IL1RL2
Format Purified
Host Species Mouse
Immunogen Partial recombinant human IL1RL2 (between amino acids 1-250) [UniProt Q9HB29]
Isotype κ
Quantity 0.025mg
Conjugate Unlabeled
Cross Reactivity Human
Regulatory Status RUO
Clone 15G9
Applications Western Blotting