Please login to your online account to display your discounted pricing

anti-IL1RL2, Clone: 15G9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP233299

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP233299

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Monoclonal or Polyclonal Monoclonal
Concentration 1.0mg/mL
Conjugate Unlabeled
Primary or Secondary Primary
Gene ID (Entrez) 8808
Gene Accession No. Q9HB29
Applications Western Blotting
Isotype κ
Regulatory Status RUO
Host Species Mouse
Immunogen Partial recombinant human IL1RL2 (between amino acids 1-250) [UniProt Q9HB29]
Quantity 0.1mg
Clone 15G9
Antigen IL1RL2
Cross Reactivity Human
Format Purified