Please login to your online account to display your discounted pricing

anti-IL1RL2, Clone: 15G9, Novus Biologicals™

$109.00 - $307.00

Specifications

Clone 15G9
Host Species Murine
Isotype κ
Immunogen Partial recombinant human IL1RL2 (between amino acids 1-250) [UniProt Q9HB29]
Antigen IL1RL2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP233299

 
novus biologicals canada
NBP233299
Unlabeled 0.1mg Each for $307.00

NBP233299SS

 
novus biologicals canada
NBP233299SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Clone 15G9
Host Species Murine
Isotype κ
Immunogen Partial recombinant human IL1RL2 (between amino acids 1-250) [UniProt Q9HB29]
Antigen IL1RL2
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Gene Accession No. Q9HB29
Gene ID (Entrez) 8808
Formulation PBS
Concentration 1.0mg/mL