Please login to your online account to display your discounted pricing

anti-Mouse IL-9, Polyclonal, R&D Systems


Manufacturer: r & d systems  RDSAF409

 View more versions of this product

Catalog No. AF409

  • / Each
 

Description & Specifications

Specifications

Antigen IL-9
Host Species Goat
Isotype IgG
Cross Reactivity Mouse
Format Affinity Purified
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL&No.x2011;9, Gln19-Pro144 (Gln19, predicted) ES: Verify IMM., Accession No. P15247
Primary or Secondary Primary
Conjugate Unlabeled
Gene ID (Entrez) 16198
Monoclonal or Polyclonal Polyclonal
Quantity 100μg
Regulatory Status RUO
Gene Accession No. P15247
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.

IL-9 Polyclonal antibody AF409 Specifically detects IL-9 in Mouse samples. It is validated for Neutralization, Western Blotting