missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Mouse IL-9, Polyclonal, R&D Systems

$447.12 - $467.98

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. P15247
Gene ID (Entrez) 16198
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AF409 R & D Systems
RDSAF409
100μg Each for $447.12
AB409NA R & D Systems
RDSAB409NA
1mg Each for $467.98
Specifications

Description

Host Species: Goat
Species Reactivity:Mouse
Immunogen:S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL&No.x2011;9, Gln19-Pro144, Accession No. P15247

Applications:

Neutralization, Western Blotting

Description & Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
P15247
16198