Please login to your online account to display your discounted pricing

anti-IL-7 R alpha/CD127, Clone: 22F3D9, Novus Biologicals™

$109.00 - $252.00

Specifications

Clone 22F3D9
Host Species Mouse
Isotype λ
Immunogen Amino acids 141-237 of human CD127 were used as the immunogen for the antibody.
Antigen IL-7 R alpha/CD127
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP222376

 
novus biologicals canada
NBP222376
Unlabeled 0.1mg Each for $252.00

NBP222376SS

 
novus biologicals canada
NBP222376SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Clone 22F3D9
Host Species Mouse
Isotype λ
Immunogen Amino acids 141-237 of human CD127 were used as the immunogen for the antibody.
Antigen IL-7 R alpha/CD127
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene Accession No. NP_002176.2
Gene ID (Entrez) 3575
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA