missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Mouse IL-7, Polyclonal, R&D Systems

$467.98 - $481.01

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. Q544C8
Gene ID (Entrez) 16196
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AF407 R & D Systems
RDSAF407
100μg Each for $481.01
AB407NA R & D Systems
RDSAB407NA
1mg Each for $467.98
Specifications

Description

Host Species: Goat
Species Reactivity:Mouse
Immunogen:E. coli-derived recombinant mouse IL-7, Glu26-Ile154, Accession No. Q544C8

Applications:

Neutralization, Western Blotting

Description & Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Q544C8
16196