Please login to your online account to display your discounted pricing

anti-IL-4, Clone: 4D9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP177536

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP177536

  • / Each
 

Description & Specifications

Specifications

Antigen IL-4
Applications Western Blot
Clone 4D9
Conjugate Unlabeled
Cross Reactivity Human
Format Size Exclusion Chromatography
Formulation PBS, azide free, filter sterilized
Host Species Murine
Immunogen Recombinant full-length human IL-4 protein.
Isotype IgG
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3565
Target Species Human