Please login to your online account to display your discounted pricing

anti-IL-4, Clone: 4D9, Novus Biologicals™

$197.00 - $384.00

Specifications

Clone 4D9
Host Species Murine
Isotype IgG
Immunogen Recombinant full-length human IL-4 protein.
Antigen IL-4
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP177536

 
novus biologicals canada
NBP177536
Unlabeled 0.1mg Each for $384.00

NBP177536SS

 
novus biologicals canada
NBP177536SS
Unlabeled 0.025mg Each for $197.00
Description & Specifications

Specifications

Clone 4D9
Host Species Murine
Isotype IgG
Immunogen Recombinant full-length human IL-4 protein.
Antigen IL-4
Monoclonal or Polyclonal Monoclonal
Format Size Exclusion Chromatography
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Gene ID (Entrez) 3565
Formulation PBS, azide free, filter sterilized