Please login to your online account to display your discounted pricing

anti-IL-33, Clone: 6H496, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP230103

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP230103

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Antigen IL-33
Isotype κ
Clone 6H496
Cross Reactivity Human
Format Purified
Applications Simple Western
Applications Western Blotting
Conjugate Unlabeled
Gene ID (Entrez) 90865
Primary or Secondary Primary
Formulation PBS with 0.05% BSA
Monoclonal or Polyclonal Monoclonal
Host Species Mouse
Concentration 0.5mg/mL
Immunogen A partial recombinant human IL-33 (amino acids 112-270) protein was used as immunogen for this antibody.
Quantity 0.1mg