Please login to your online account to display your discounted pricing

anti-IL-33, Clone: 6H496, Novus Biologicals™

$139.00 - $332.66

Specifications

Clone 6H496
Host Species Mouse
Isotype κ
Immunogen A partial recombinant human IL-33 (amino acids 112-270) protein was used as immunogen for this antibody.
Antigen IL-33
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP230103SS

 
novus biologicals canada
NBP230103SS
Unlabeled 0.025mg Each for $139.00

NBP230103

 
novus biologicals canada
NBP230103
Unlabeled 0.1mg Each for $332.66
Description & Specifications

Specifications

Clone 6H496
Host Species Mouse
Isotype κ
Immunogen A partial recombinant human IL-33 (amino acids 112-270) protein was used as immunogen for this antibody.
Antigen IL-33
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Simple Western
Applications Western Blotting
Gene ID (Entrez) 90865
Formulation PBS with 0.05% BSA
Concentration 0.5mg/mL