Please login to your online account to display your discounted pricing

anti-IL-33, Clone: 6H496, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP230103SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP230103SS

  • / Each
 

Description & Specifications

Specifications

Formulation PBS with 0.05% BSA
Monoclonal or Polyclonal Monoclonal
Applications Simple Western
Applications Western Blotting
Conjugate Unlabeled
Primary or Secondary Primary
Format Purified
Clone 6H496
Gene ID (Entrez) 90865
Isotype κ
Immunogen A partial recombinant human IL-33 (amino acids 112-270) protein was used as immunogen for this antibody.
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Quantity 0.025mg
Antigen IL-33
Concentration 0.5mg/mL