Please login to your online account to display your discounted pricing

anti-IL-33, Clone: 1C5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP231145SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP231145SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. O95760
Primary or Secondary Primary
Concentration 1.0mg/mL
Formulation PBS
Cross Reactivity Human
Cross Reactivity Mouse
Monoclonal or Polyclonal Monoclonal
Quantity 0.025mg
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Synthetic peptide corresponding to an internal sequence of the human IL33 protein (between amino acids 100-270) [UniProt O95760]
Isotype κ
Format Purified
Clone 1C5
Regulatory Status RUO
Host Species Mouse
Conjugate Unlabeled
Antigen IL-33
Gene ID (Entrez) 90865