Please login to your online account to display your discounted pricing

anti-IL-33, Clone: 1C5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP231145

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP231145

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Primary or Secondary Primary
Gene ID (Entrez) 90865
Concentration 1.0mg/mL
Conjugate Unlabeled
Gene Accession No. O95760
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Cross Reactivity Mouse
Isotype κ
Clone 1C5
Format Purified
Host Species Mouse
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Synthetic peptide corresponding to an internal sequence of the human IL33 protein (between amino acids 100-270) [UniProt O95760]
Quantity 0.1mg
Regulatory Status RUO
Antigen IL-33