Please login to your online account to display your discounted pricing

anti-IL-33, Clone: 1C5, Novus Biologicals™

$109.00 - $329.00

Specifications

Clone 1C5
Host Species Mouse
Isotype κ
Immunogen Synthetic peptide corresponding to an internal sequence of the human IL33 protein (between amino acids 100-270) [UniProt O95760]
Antigen IL-33
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP231145

 
novus biologicals canada
NBP231145
Unlabeled 0.1mg Each for $329.00

NBP231145SS

 
novus biologicals canada
NBP231145SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Clone 1C5
Host Species Mouse
Isotype κ
Immunogen Synthetic peptide corresponding to an internal sequence of the human IL33 protein (between amino acids 100-270) [UniProt O95760]
Antigen IL-33
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene Accession No. O95760
Gene ID (Entrez) 90865
Formulation PBS
Concentration 1.0mg/mL