Please login to your online account to display your discounted pricing

anti-IL-3 R alpha/CD123, Clone: 18N6B12, Novus Biologicals™

$109.00 - $255.80

Specifications

Clone 18N6B12
Host Species Murine
Immunogen RERVYEFLSAWSTPQRFEC (aa275-293).
Antigen IL-3 R alpha/CD123
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227225

 
novus biologicals canada
NBP227225
Unlabeled 0.1mg Each for $255.80

NBP227225SS

 
novus biologicals canada
NBP227225SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Clone 18N6B12
Host Species Murine
Immunogen RERVYEFLSAWSTPQRFEC (aa275-293).
Antigen IL-3 R alpha/CD123
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Western Blot
Gene Accession No. P26951
Gene ID (Entrez) 3563
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Concentration 0.5mg/mL