Please login to your online account to display your discounted pricing

anti-Mouse IL-3, Polyclonal, R&D Systems


Manufacturer: r & d systems  RDSAB403NA

 View more versions of this product

Catalog No. AB403NA

  • / Each
 

Description & Specifications

Specifications

Antigen IL-3
Host Species Goat
Immunogen E. coli-derived recombinant mouse IL-3, Asp33-Cys166, Accession No. P01586
Quantity 1mg
Cross Reactivity Mouse
Primary or Secondary Primary
Conjugate Unlabeled
Gene ID (Entrez) 16187
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO
Format Affinity Purified
Isotype IgG
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. P01586