missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Mouse IL-3, Polyclonal, R&D Systems

$454.80 - $454.80

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. P01586
Gene ID (Entrez) 16187
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AB403NA R & D Systems
RDSAB403NA
1mg Each for $454.80
AF403NA R & D Systems
RDSAF403NA
100μg Each for $454.80
Specifications

Description

Host Species: Goat
Species Reactivity:Mouse
Immunogen:E. coli-derived recombinant mouse IL-3, Asp33-Cys166, Accession No. P01586

Applications:

Neutralization, Western Blotting

Description & Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
P01586
16187