Please login to your online account to display your discounted pricing

anti-Mouse IL-3, Polyclonal, R&D Systems


Manufacturer: r & d systems  RDSAF403NA

 View more versions of this product

Catalog No. AF403NA

  • / Each
 

Description & Specifications

Specifications

Antigen IL-3
Conjugate Unlabeled
Cross Reactivity Mouse
Format Affinity Purified
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. P01586
Host Species Goat
Immunogen E. coli-derived recombinant mouse IL-3, Asp33-Cys166, Accession No. P01586
Isotype IgG
Quantity 100μg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 16187
Target Species Murine

IL-3 Polyclonal antibody AF403SP Specifically detects IL-3 in Mouse samples. It is validated for Neutralization, Western Blotting