Please login to your online account to display your discounted pricing

anti-IL-27 R alpha/WSX-1/TCCR, Clone: 34N4G11, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP219015SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP219015SS

  • / Each
 

Description & Specifications

Specifications

Antigen IL-27 R alpha/WSX-1/TCCR
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Clone 34N4G11
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS containing 0.05% BSA
Gene Accession No. Q6UWB1
Host Species Murine
Immunogen A portion of amino acids 200-250 of human IL-27R (WSX-1) was used as the immunogen for this antibody.
Isotype κ
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 9466
Test Specificity IL-27R (WSX-1) is 65-70 kDa protein and is highly expressed in lymphoid tissues such as spleen, lymph nodes and peripheral blood leukocytes including CD4+ and CD8+ T cells, monocytes, DCs and NK cells.
Target Species Human