Please login to your online account to display your discounted pricing

anti-IL-27/IL-35 EBI3 Subunit, Clone: 42N12F7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227357

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227357

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 10148
Conjugate Unlabeled
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL
Monoclonal or Polyclonal Monoclonal
Gene Accession No. Q14213
Primary or Secondary Primary
Cross Reactivity Human
Antigen IL-27/IL-35 EBI3 Subunit
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Full-length recombinant human EBI3 protein full length recombinant human EBI3 (cat No. 100-162)
Quantity 0.1mg
Host Species Mouse
Clone 42N12F7
Format Purified
Regulatory Status RUO