Please login to your online account to display your discounted pricing

anti-IL-27/IL-35 EBI3 Subunit, Clone: 42N12F7, Novus Biologicals™

$139.00 - $307.00

Specifications

Clone 42N12F7
Host Species Murine
Immunogen Full-length recombinant human EBI3 protein full length recombinant human EBI3 (cat No. 100-162)
Antigen IL-27/IL-35 EBI3 Subunit
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227357

 
novus biologicals canada
NBP227357
Unlabeled 0.1mg Each for $307.00

NBP227357SS

 
novus biologicals canada
NBP227357SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 42N12F7
Host Species Murine
Immunogen Full-length recombinant human EBI3 protein full length recombinant human EBI3 (cat No. 100-162)
Antigen IL-27/IL-35 EBI3 Subunit
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene Accession No. Q14213
Gene ID (Entrez) 10148
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL