Please login to your online account to display your discounted pricing

anti-IL-27/IL-35 EBI3 Subunit, Clone: 42N12F7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227357SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227357SS

  • / Each
 

Description & Specifications

Specifications

Antigen IL-27/IL-35 EBI3 Subunit
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Clone 42N12F7
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Gene Accession No. Q14213
Host Species Murine
Immunogen Full-length recombinant human EBI3 protein full length recombinant human EBI3 (cat No. 100-162)
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 10148
Target Species Human