Please login to your online account to display your discounted pricing

anti-IL-23 R, Clone: 15N6C6, Novus Biologicals™

$109.00 - $255.80

Specifications

Clone 15N6C6
Host Species Murine
Isotype κ
Immunogen A synthetic peptide corresponding to amino acids 288-315 of hIL-23 receptor was used as immunogen for this antibody.
Antigen IL-23 R
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227091

 
novus biologicals canada
NBP227091
Unlabeled 0.1mg Each for $255.80

NBP227091SS

 
novus biologicals canada
NBP227091SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Clone 15N6C6
Host Species Murine
Isotype κ
Immunogen A synthetic peptide corresponding to amino acids 288-315 of hIL-23 receptor was used as immunogen for this antibody.
Antigen IL-23 R
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in Human, Chimpanzee and Orangutan, 89% in Mouse, Pig, Dog and Rhesus Monkey, and 84% homologous in Rat, Horse and Bovine. There are multiple isoforms of IL-23 receptor. Researchers are encouraged to use a tool like BLAST to determine the suitability of this antibody for their testing.
Gene Accession No. Q5VWk5
Gene ID (Entrez) 149233
Formulation 100μg antibody in 200μL PBS with 0.05% BSA.
Concentration 0.5mg/mL