Please login to your online account to display your discounted pricing

anti-IL-22, Clone: 8F7H11.2E11, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227360

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227360

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 50616
Concentration 0.5mg/mL
Primary or Secondary Primary
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Conjugate Unlabeled
Gene Accession No. Q9GZX6
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Applications Western Blotting
Antigen IL-22
Clone 8F7H11.2E11
Quantity 0.1mg
Immunogen MAPISSHCRL DkSNFQQPYI TNRTFMLAkE ASLADNNTDV RLIGEkLFHG VSMSERCYLM kQVLNFTLEE VLFPQSDRFQ PYMQEVVPFL ARLSNRLSTC HIEGDDLHIQ RNVQkLkDTV kkLGESGEIk AIGELDLLFM SLRNACI
Host Species Mouse
Format Purified
Cross Reactivity Human