Please login to your online account to display your discounted pricing

anti-IL-22, Clone: 8F7H11.2E11, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227360SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227360SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q9GZX6
Primary or Secondary Primary
Concentration 0.5mg/mL
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Format Purified
Antigen IL-22
Immunogen MAPISSHCRL DkSNFQQPYI TNRTFMLAkE ASLADNNTDV RLIGEkLFHG VSMSERCYLM kQVLNFTLEE VLFPQSDRFQ PYMQEVVPFL ARLSNRLSTC HIEGDDLHIQ RNVQkLkDTV kkLGESGEIk AIGELDLLFM SLRNACI
Clone 8F7H11.2E11
Regulatory Status RUO
Host Species Mouse
Applications Western Blotting
Quantity 0.025mg
Gene ID (Entrez) 50616