Please login to your online account to display your discounted pricing

anti-IL-22, Clone: 8F7H11.2B9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227320SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227320SS

  • / Each
 

Description & Specifications

Specifications

Antigen IL-22
Applications Western Blot
Clone 8F7H11.2B9
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Gene Accession No. Q9GZX6
Host Species Murine
Immunogen Full-length recombinant human IL-22 protein was used as immunogen for this antibody.
Isotype κ
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 50616
Target Species Human