Please login to your online account to display your discounted pricing

anti-IL-22, Clone: 8F7H11.2B9, Novus Biologicals™

$109.00 - $255.80

Specifications

Clone 8F7H11.2B9
Host Species Murine
Isotype κ
Immunogen Full-length recombinant human IL-22 protein was used as immunogen for this antibody.
Antigen IL-22
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227320SS

 
novus biologicals canada
NBP227320SS
Unlabeled 0.025mg Each for $109.00

NBP227320

 
novus biologicals canada
NBP227320
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Clone 8F7H11.2B9
Host Species Murine
Isotype κ
Immunogen Full-length recombinant human IL-22 protein was used as immunogen for this antibody.
Antigen IL-22
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Gene Accession No. Q9GZX6
Gene ID (Entrez) 50616
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Concentration 0.5mg/mL