Please login to your online account to display your discounted pricing

anti-IL-22, Clone: 8F10C10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227321

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227321

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 50616
Gene Accession No. Q9GZX6
Format Purified
Host Species Mouse
Clone 8F10C10
Regulatory Status RUO
Cross Reactivity Human
Primary or Secondary Primary
Immunogen MAPISSHCRL DkSNFQQPYI TNRTFMLAkE ASLADNNTDV RLIGEkLFHG VSMSERCYLM kQVLNFTLEE VLFPQSDRFQ PYMQEVVPFL ARLSNRLSTC HIEGDDLHIQ RNVQkLkDTV kkLGESGEIk AIGELDLLFM SLRNACI
Concentration 0.5mg/mL
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Isotype κ
Quantity 0.1mg
Antigen IL-22
Applications Western Blotting