Please login to your online account to display your discounted pricing

anti-IL-22, Clone: 8F10C10, Novus Biologicals™

$109.00 - $255.80

Specifications

Clone 8F10C10
Host Species Murine
Isotype κ
Immunogen MAPISSHCRL DkSNFQQPYI TNRTFMLAkE ASLADNNTDV RLIGEkLFHG VSMSERCYLM kQVLNFTLEE VLFPQSDRFQ PYMQEVVPFL ARLSNRLSTC HIEGDDLHIQ RNVQkLkDTV kkLGESGEIk AIGELDLLFM SLRNACI
Antigen IL-22
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227321

 
novus biologicals canada
NBP227321
Unlabeled 0.1mg Each for $255.80

NBP227321SS

 
novus biologicals canada
NBP227321SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Clone 8F10C10
Host Species Murine
Isotype κ
Immunogen MAPISSHCRL DkSNFQQPYI TNRTFMLAkE ASLADNNTDV RLIGEkLFHG VSMSERCYLM kQVLNFTLEE VLFPQSDRFQ PYMQEVVPFL ARLSNRLSTC HIEGDDLHIQ RNVQkLkDTV kkLGESGEIk AIGELDLLFM SLRNACI
Antigen IL-22
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Gene Accession No. Q9GZX6
Gene ID (Entrez) 50616
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Concentration 0.5mg/mL